accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3(Z)

CAS RN 2733-88-2

IUPAC Name: methyl (15Z)-tetracos-15-enoate
Synonyms: methyl (15Z)-tetracos-15-enoate
Molecular Weight (g/mol): 380.66
Molecular Formula: C25H48O2
InChi Key: AINIZSBLAFHZCP-KHPPLWFESA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCCCCCCC(=O)OC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results