Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl nervonate, 99%, analytical standard for GC

Catalog No. AC460375000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460375000 500 mg
AC460371000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460375000 Supplier Thermo Scientific Chemicals Supplier No. 460375000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2733-88-2 | C25H48O2 | 380.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2733-88-2
Molecular Formula C25H48O2
Molecular Weight (g/mol) 380.65
InChI Key AINIZSBLAFHZCP-KHPPLWFESA-N
Synonym nervonic acid methyl ester, methyl nervonate, methyl z-tetracos-15-enoate, methyl cis-15-tetracosenoate, selacholeic acid methyl ester, 15-tetracosenoic acid, methyl ester, z, methyl nervonate c24:1, z-methyl tetracos-15-enoate, methyl 15z tetracos-15-enoate, methyl 15z-15-tetracosenoate #
PubChem CID 5364841
IUPAC Name methyl (Z)-tetracos-15-enoate
SMILES CCCCCCCCC=CCCCCCCCCCCCCCC(=O)OC

Specifications

Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)7CHCH(CH2)13COOCH3
Quantity 500 mg
Formula Weight 380.65
Percent Purity 99%
Chemical Name or Material Methyl nervonate
EINECSNumber 220-352-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.