accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27333-47-7

IUPAC Name: 1,3,3-trimethyl-1,3-dihydrospiro[indole-2,3'-naphtho[2,1-b][1,4]oxazine]
Synonyms: 1,3,3-trimethylspiro[indole-2,3'-naphtho[2,1-b][1,4]oxazine]
Molecular Weight (g/mol): 328.42
Molecular Formula: C22H20N2O
InChi Key: CQTRKDFIQFOAQV-UHFFFAOYNA-N
SMILES: CN1C2=CC=CC=C2C(C)(C)C11OC2=CC=C3C=CC=CC3=C2N=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results