Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,3-Trimethylindolinonaphthospirooxazine 98.0+%, TCI America™
SDP

Catalog No. T12591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12591G Supplier TCI America Supplier No. T12591G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27333-47-7
Molecular Formula C22H20N2O
Molecular Weight (g/mol) 328.415
MDL Number MFCD00013318
InChI Key CQTRKDFIQFOAQV-UHFFFAOYSA-N
Synonym 1,3,3-Trimethylspiro[indoline-2,3′C-[3H]naphth[2,1-b][1,4]oxazine]
PubChem CID 594662
IUPAC Name 1',3',3'-trimethylspiro[benzo[f][1,4]benzoxazine-3,2'-indole]
SMILES CC1(C2=CC=CC=C2N(C13C=NC4=C(O3)C=CC5=CC=CC=C54)C)C

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 1 g
Formula Weight 328.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,3-Trimethylindolinonaphthospirooxazine [Photochromic Compound]
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.