accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3H2N

CAS RN 2734-70-5

IUPAC Name: 2,6-dimethoxyaniline
Synonyms: 2,6-dimethoxyaniline
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: HQBJSEKQNRSDAZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=C1N
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)