Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxyaniline, 96%

Catalog No. AC448790050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448790050 5 g Glass bottle
AC448790010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448790050 Supplier Thermo Scientific Chemicals Supplier No. 448790050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2734-70-5 | C8H11NO2 | 153.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2734-70-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
InChI Key HQBJSEKQNRSDAZ-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dimethoxy, 2,6-dimethoxyphenylamine, 2,6-dimethoxybenzenamine, 2,6-dimethoxy-phenylamine, buttpark 33\04-69, 2 6-dimethoxyaniline, pubchem4419, 2,6-dimethoxy aniline, acmc-1cnwd, ksc497k1h
PubChem CID 95940
IUPAC Name 2,6-dimethoxyaniline
SMILES COC1=C(C(=CC=C1)OC)N

Specifications

Melting Point 76°C to 78°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3NH2
Quantity 5 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 153.18
Percent Purity 96%
Chemical Name or Material 2, 6-Dimethoxyaniline
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause damage to organs through p
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.