accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NO

CAS RN 27387-31-1

IUPAC Name: 9-methyl-2,3,4,9-tetrahydro-1H-carbazol-4-one
Synonyms: 9-methyl-2,3-dihydro-1H-carbazol-4-one
Molecular Weight (g/mol): 199.25
Molecular Formula: C13H13NO
InChi Key: HHJUJCWZKJMCLC-UHFFFAOYSA-N
SMILES: CN1C2=C(C3=CC=CC=C13)C(=O)CCC2
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)