Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydro-9-methylcarbazol-4-one 98.0+%, TCI America™
SDP

Catalog No. T29425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29425G 5 g
T294225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29425G Supplier TCI America Supplier No. T29425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27387-31-1
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.253
MDL Number MFCD00173748
InChI Key HHJUJCWZKJMCLC-UHFFFAOYSA-N
Synonym 1,2,3,4-Tetrahydro-9-methyl-4-oxocarbazole
PubChem CID 598875
IUPAC Name 9-methyl-2,3-dihydro-1H-carbazol-4-one
SMILES CN1C2=C(C(=O)CCC2)C3=CC=CC=C31

Specifications

Melting Point 201°C
Color White-Yellow
Quantity 5 g
Formula Weight 199.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Tetrahydro-9-methylcarbazol-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.