accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 274-40-8

IUPAC Name: indolizine
Synonyms: indolizine pyrrocoline
Molecular Weight (g/mol): 117.15
Molecular Formula: C8H7N
InChi Key: HOBCFUWDNJPFHB-UHFFFAOYSA-N
SMILES: C1=CN2C=CC=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results