Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrolo[1,2-a]pyridine, TCI America™
SDP

Catalog No. P19431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19431G Supplier TCI America Supplier No. P19431G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 274-40-8
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.151
MDL Number MFCD00030189
InChI Key HOBCFUWDNJPFHB-UHFFFAOYSA-N
Synonym 4-Azaindene, Indolizine
PubChem CID 9230
ChEBI CHEBI:35583
IUPAC Name indolizine
SMILES C1=CC2=CC=CN2C=C1

Specifications

Melting Point 74°C
Color White-Yellow
Boiling Point 202°C
Quantity 1 g
Formula Weight 117.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrrolo[1,2-a]pyridine
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.