accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNN

CAS RN 274-95-3

IUPAC Name: imidazo[1,2-a]pyrimidine
Synonyms: imidazo[1,2-a]pyrimidine
Molecular Weight (g/mol): 119.13
Molecular Formula: C6H5N3
InChi Key: INSWZAQOISIYDT-UHFFFAOYSA-N
SMILES: C1=CN2C=CC=NC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results