Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazo[1,2-a]pyrimidine 98.0+%, TCI America™
SDP

Catalog No. I07985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07985G 5 g
I07981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07985G Supplier TCI America Supplier No. I07985G

Chemical Identifiers

CAS 274-95-3
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.13
MDL Number MFCD06245377
InChI Key INSWZAQOISIYDT-UHFFFAOYSA-N
PubChem CID 577018
IUPAC Name imidazo[1,2-a]pyrimidine
SMILES C1=CN2C=CC=NC2=N1

Specifications

Melting Point 127°C
Quantity 5 g
Formula Weight 119.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Imidazo[1,2-a]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.