accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27425-92-9

IUPAC Name: 2,5,8,11,14,17,20,23,26,29-decaoxahentriacontan-31-ol
Synonyms: 2,5,8,11,14,17,20,23,26,29-decaoxahentriacontan-31-ol
Molecular Weight (g/mol): 472.57
Molecular Formula: C21H44O11
InChi Key: YMBLPWKGRIGDBP-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results