Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decaethylene Glycol Monomethyl Ether 93.0+%, TCI America™
SDP

Catalog No. D2903100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2903100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2903100MG Supplier TCI America Supplier No. D2903100MG

Chemical Identifiers

CAS 27425-92-9
Molecular Formula C21H44O11
Molecular Weight (g/mol) 472.57
MDL Number MFCD06797087
InChI Key YMBLPWKGRIGDBP-UHFFFAOYSA-N
Synonym mPEG10-Alcohol
PubChem CID 14550547
IUPAC Name 2,5,8,11,14,17,20,23,26,29-decaoxahentriacontan-31-ol
SMILES COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Melting Point 19°C
Color Yellow
Quantity 100 mg
Formula Weight 472.57
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Decaethylene Glycol Monomethyl Ether
EINECSNumber (7)-0097
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.