accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OOOOH3CCH3

CAS RN 2744-50-5

IUPAC Name: 3,9-bis(2-methylpropyl) perylene-3,9-dicarboxylate
Synonyms: 3,9-bis(2-methylpropyl) perylene-3,9-dicarboxylate
Molecular Weight (g/mol): 452.55
Molecular Formula: C30H28O4
InChi Key: YLNJGHNUXCVDIX-UHFFFAOYSA-N
SMILES: CC(C)COC(=O)C1=CC=C2C3=CC=CC4=C(C=CC(C5=CC=CC1=C25)=C34)C(=O)OCC(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results