Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisobutyl Perylenedicarboxylate (mixture of regioisomers) 98.0+%, TCI America™
SDP

Catalog No. P11975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11975G 5 g
P119725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11975G Supplier TCI America Supplier No. P11975G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2744-50-5
Molecular Formula C30H28O4
Molecular Weight (g/mol) 452.55
MDL Number MFCD00191681
InChI Key YLNJGHNUXCVDIX-UHFFFAOYSA-N
Synonym Perylenedicarboxylic Acid Diisobutyl Ester
PubChem CID 75973
IUPAC Name bis(2-methylpropyl) perylene-3,9-dicarboxylate
SMILES CC(C)COC(=O)C1=CC=C2C3=C4C(=CC=C3)C(=CC=C4C5=C2C1=CC=C5)C(=O)OCC(C)C

Specifications

Quantity 5 g
Formula Weight 452.55
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Diisobutyl Perylenedicarboxylate (mixture of regioisomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.