accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27458-90-8

IUPAC Name: 2-methyl-2-[(2-methylundecan-2-yl)disulfanyl]undecane
Synonyms: 2-methyl-2-[(2-methylundecan-2-yl)disulfanyl]undecane
Molecular Weight (g/mol): 402.78
Molecular Formula: C24H50S2
InChi Key: LEDIWWJKWAMGLD-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(C)(C)SSC(C)(C)CCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results