Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-dodecyl Disulfide (mixture of isomers), TCI America™
SDP

Catalog No. D044725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D044725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D044725G Supplier TCI America Supplier No. D044725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27458-90-8
Molecular Formula C24H50S2
Molecular Weight (g/mol) 402.784
MDL Number MFCD00059944
InChI Key LEDIWWJKWAMGLD-UHFFFAOYSA-N
Synonym tert-Dodecyl Disulfide
PubChem CID 117981
ChEBI CHEBI:84283
IUPAC Name 2-methyl-2-(2-methylundecan-2-yldisulfanyl)undecane
SMILES CCCCCCCCCC(C)(C)SSC(C)(C)CCCCCCCCC

Specifications

Color Yellow
Boiling Point 321°C
Quantity 25 g
Formula Weight 402.78
Physical Form Liquid
Chemical Name or Material Di-tert-dodecyl Disulfide (mixture of isomers)
EINECSNumber (2)-0473&(2)-0475
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.