accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27465-51-6

IUPAC Name: 1-(4-ethylphenyl)propan-1-one
Synonyms: 1-(4-ethylphenyl)propan-1-one
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: VGQRIILEZYZAOE-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(CC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results