Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Ethylpropiophenone 96.0+%, TCI America™
SDP

Catalog No. E046125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E046125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E046125ML Supplier TCI America Supplier No. E046125ML

missing translation for 'validMainframeMsg'

Specifications

CAS 27465-51-6
Color Colorless
Boiling Point 246°C
Molecular Formula C11H14O
Quantity 25 mL
MDL Number MFCD00191646
Synonym Ethyl 4-Ethylphenyl Ketone
InChI Key VGQRIILEZYZAOE-UHFFFAOYSA-N
SMILES CCC1=CC=C(C=C1)C(=O)CC
IUPAC Name 1-(4-ethylphenyl)propan-1-one
Molecular Weight (g/mol) 162.232
PubChem CID 583750
Formula Weight 162.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Ethylpropiophenone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.