accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNH

CAS RN 274694-98-3

IUPAC Name: 11-methyl-11,21-diazatetracyclo[12.7.0.0³,⁸.0¹⁵,²⁰]henicosa-1(14),3,5,7,15,17,19-heptaene
Synonyms: 11-methyl-11,21-diazatetracyclo[12.7.0.0³,⁸.0¹⁵,²⁰]henicosa-1(14),3,5,7,15,17,19-heptaene
Molecular Weight (g/mol): 290.41
Molecular Formula: C20H22N2
InChi Key: YEWGIGCYIAMFMA-UHFFFAOYSA-N
SMILES: CN1CCC2=C(CC3=CC=CC=C3CC1)NC1=CC=CC=C21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results