accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27473-62-7

IUPAC Name: 2-amino-2,3-dihydro-1H-indene-2-carboxylic acid
Synonyms: 2-amino-1,3-dihydroindene-2-carboxylic acid
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: UHQFXIWMAQOCAN-UHFFFAOYSA-N
SMILES: NC1(CC2=CC=CC=C2C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results