Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-2-indanecarboxylic Acid, Thermo Scientific™

Catalog No. AC338182500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC338182500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC338182500 Supplier Thermo Scientific Chemicals Supplier No. 338182500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27473-62-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD03419294
InChI Key UHQFXIWMAQOCAN-UHFFFAOYSA-N
Synonym 2-amino-2-indancarboxylic acid, 2-amino-2,3-dihydro-1h-indene-2-carboxylic acid, 2-aminoindane-2-carboxylic acid, 2-aminoindan-2-carboxylic acid, 2-aminoindan-2-carboxylicacid, 2-amino-indan-2-carboxylic acid, +/--2-aminoindane-2-carboxylic acid, 1h-indene-2-carboxylic acid, 2-amino-2,3-dihydro, 1h-indene-2-carboxylicacid, 2-amino-2,3-dihydro, 2-amino-2-indancarboxylicacid
PubChem CID 250936
IUPAC Name 2-amino-2,3-dihydro-1H-indene-2-carboxylic acid
SMILES NC1(CC2=CC=CC=C2C1)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 177.2
Chemical Name or Material 2-Amino-2-indanecarboxylic acid
Hazard Category HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.