accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNOCH3HONHOO

CAS RN 27482-74-2

IUPAC Name: ethyl 2-(2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanamido)acetate
Synonyms: ethyl 2-(2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanamido)acetate
Molecular Weight (g/mol): 338.36
Molecular Formula: C16H22N2O6
InChi Key: HJLLTKBIYMYCMV-UHFFFAOYNA-N
SMILES: CCOC(=O)CNC(=O)C(NC(=O)OCC1=CC=CC=C1)C(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results