CAS RN 27489-33-4
IUPAC Name:
2-methanesulfonylphenol
Synonyms:
2-methanesulfonylphenol
Molecular Weight (g/mol):
172.2
Molecular Formula:
C7H8O3S
InChi Key:
ZWTZHHHSRDUVRT-UHFFFAOYSA-N
SMILES:
CS(=O)(=O)C1=CC=CC=C1O
1
–
1
of
1
results
1
Promotions Available
2-(Methylsulfonyl)phenol, 98%
2-(Methylsulfonyl)phenol, 98%
CAS: 27489-33-4 Molecular Formula: C7H8O3S Molecular Weight (g/mol): 172.2 InChI Key: ZWTZHHHSRDUVRT-UHFFFAOYSA-N PubChem CID: 598998 IUPAC Name: 2-methylsulfonylphenol SMILES: CS(=O)(=O)C1=CC=CC=C1O