CAS RN 2752-65-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Gambogic Acid, Garcinia hanburyi
A cell-permeable caspase activator and an apoptosis inducer that was originally isolated from gamboge resin for its antimicrobial properties.