Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Gambogic Acid, Garcinia hanburyi

Catalog No. 34570125MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-570-125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-570-125MG Supplier MilliporeSigma™ Supplier No. 34570125MG
Add to Cart
Add to Cart

A cell-permeable caspase activator and an apoptosis inducer that was originally isolated from gamboge resin for its antimicrobial properties.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Description 95% by HPLC
Target Proteins Caspase activator and an apoptosis inducer
CAS 2752-65-0
Molecular Formula C38H44O8
Formula Weight 628.8g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Amorphous Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.