accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27530-80-9

H3COPHOHOONH2OHO

CAS RN 27530-80-9

IUPAC Name: 2-amino-3-(phosphonooxy)butanoic acid
Synonyms: phosphonothreonine
Molecular Weight (g/mol): 199.1
Molecular Formula: C4H10NO6P
InChi Key: USRGIUJOYOXOQJ-UHFFFAOYNA-N
SMILES: CC(OP(O)(O)=O)C(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)