Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Phospho-DL-threonine, TCI America™
SDP

Catalog No. P02551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P02551G Supplier TCI America Supplier No. P02551G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27530-80-9
Molecular Formula C4H10NO6P
Molecular Weight (g/mol) 199.099
MDL Number MFCD00135662
InChI Key USRGIUJOYOXOQJ-STYMGOOTSA-N
PubChem CID 133126436
IUPAC Name (3R)-2-amino-3-phosphonooxybutanoic acid
SMILES CC(C(C(=O)O)N)OP(=O)(O)O

Specifications

Melting Point 184°C
Color White
Quantity 1 g
Formula Weight 199.10
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Phospho-DL-threonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.