accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOO

CAS RN 2754-41-8

IUPAC Name: 5,11-dioxatricyclo[7.3.0.0³,⁷]dodecane-4,6,10,12-tetrone
Synonyms: 5,11-dioxatricyclo[7.3.0.0³,⁷]dodecane-4,6,10,12-tetrone
Molecular Weight (g/mol): 224.17
Molecular Formula: C10H8O6
InChi Key: LJMPOXUWPWEILS-UHFFFAOYNA-N
SMILES: O=C1OC(=O)C2CC3C(CC12)C(=O)OC3=O
Percent Purity 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results