Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4,5-Cyclohexanetetracarboxylic Dianhydride (purified by sublimation) 98.0+%, TCI America™
SDP

Catalog No. C29195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29195G 5 g
C29191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29195G Supplier TCI America Supplier No. C29195G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2754-41-8 | C10H8O6 | 224.17 g/mol

Chemical Identifiers

CAS 2754-41-8
Molecular Formula C10H8O6
Molecular Weight (g/mol) 224.17
MDL Number MFCD13704912
InChI Key LJMPOXUWPWEILS-UHFFFAOYNA-N
PubChem CID 14453667
IUPAC Name 5,11-dioxatricyclo[7.3.0.0³,⁷]dodecane-4,6,10,12-tetrone
SMILES O=C1OC(=O)C2CC3C(CC12)C(=O)OC3=O

Specifications

Melting Point 240°C
Color White
Quantity 5 g
Formula Weight 224.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4,5-Cyclohexanetetracarboxylic Dianhydride (purified by sublimation)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.