CAS RN 27563-67-3
IUPAC Name:
N-methyldodecanamide
Synonyms:
N-methyldodecanamide
Molecular Weight (g/mol):
213.37
Molecular Formula:
C13H27NO
InChi Key:
APWSJINSLHHRPD-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC(=O)NC
1
–
1
of
1
results
1
N-Methyldodecanamide 98.0+%, TCI America™
CAS: 27563-67-3 Molecular Formula: C13H27NO Molecular Weight (g/mol): 213.365 MDL Number: MFCD00051780 InChI Key: APWSJINSLHHRPD-UHFFFAOYSA-N Synonym: N-Methyllauramide, N-Dodecanoylmethylamine, N-Lauroylmethylamine PubChem CID: 141362 IUPAC Name: N-methyldodecanamide SMILES: CCCCCCCCCCCC(=O)NC