Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyldodecanamide 98.0+%, TCI America™
SDP

Catalog No. M24525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24525G Supplier TCI America Supplier No. M24525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27563-67-3
Molecular Formula C13H27NO
Molecular Weight (g/mol) 213.365
MDL Number MFCD00051780
InChI Key APWSJINSLHHRPD-UHFFFAOYSA-N
Synonym N-Methyllauramide, N-Dodecanoylmethylamine, N-Lauroylmethylamine
PubChem CID 141362
IUPAC Name N-methyldodecanamide
SMILES CCCCCCCCCCCC(=O)NC

Specifications

Melting Point 70°C
Color White
Quantity 5 g
Formula Weight 213.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyldodecanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.