accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2757-28-0

IUPAC Name: tris(6-methylheptyl)amine
Synonyms: tris(6-methylheptyl)amine
Molecular Weight (g/mol): 353.68
Molecular Formula: C24H51N
InChi Key: YKGBNAGNNUEZQC-UHFFFAOYSA-N
SMILES: CC(C)CCCCCN(CCCCCC(C)C)CCCCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results