Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisooctylamine (Technical), MP Biomedicals

Catalog No. ICN219373 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219373 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219373 Supplier MP Biomedicals, Inc Supplier No. 0521937380

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2757-28-0
Molecular Formula C24H51N
Molecular Weight (g/mol) 353.68
MDL Number MFCD00011694
InChI Key YKGBNAGNNUEZQC-UHFFFAOYSA-N
Synonym triisooctylamine, tris 6-methylheptyl amine, unii-r555wmx5uk, triheptylamine, 6,6',6-trimethyl, isooctanamine, n,n-diisooctyl, 1-heptanamine, 6-methyl-n,n-bis 6-methylheptyl, r555wmx5uk, 6-methyl-n,n-bis 6-methylheptyl heptan-1-amine, 1-heptanamine,6-methyl-n,n-bis 6-methylheptyl, 6,6',6-trimethyltriheptylamine
PubChem CID 75991
IUPAC Name tris(6-methylheptyl)amine
SMILES CC(C)CCCCCN(CCCCCC(C)C)CCCCCC(C)C

Specifications

Quantity 100 g
Formula Weight 353.68
Chemical Name or Material Triisooctylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.