accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2N

CAS RN 27578-60-5

IUPAC Name: 2-(piperidin-1-yl)ethan-1-amine
Synonyms: 2-(piperidin-1-yl)ethanamine
Molecular Weight (g/mol): 128.22
Molecular Formula: C7H16N2
InChi Key: CJNRGSHEMCMUOE-UHFFFAOYSA-N
SMILES: NCCN1CCCCC1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)