accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOFOFFFOFFFOFFFFFFFFFFFFFFFF

CAS RN 27639-98-1

IUPAC Name: 2,3,3,3-tetrafluoro-2-{1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy}propanoyl fluoride
Synonyms: 2,3,3,3-tetrafluoro-2-{1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy}propanoyl fluoride
Molecular Weight (g/mol): 664.09
Molecular Formula: C12F24O4
InChi Key: IIYQDKPSLAINNX-UHFFFAOYNA-N
SMILES: FC(=O)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results