Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride, 97%

Catalog No. AAL1661414
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1661414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1661414 Supplier Thermo Scientific Chemicals Supplier No. L1661414
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 27639-98-1 | C12F24O4 | 664.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27639-98-1
Molecular Formula C12F24O4
Molecular Weight (g/mol) 664.09
MDL Number MFCD00084743
InChI Key IIYQDKPSLAINNX-UHFFFAOYSA-N
Synonym perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-heptafluoropropoxy propoxy propoxy, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy propanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy, propanoyl fluoride,2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy, acmc-20ajy8, 3,6,9-trioxadodecanoyl fluoride, tetradecafluoro-2,5,8-tris trifluoromethyl
PubChem CID 119616
IUPAC Name 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]propanoyl fluoride
SMILES C(=O)(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F)(F)F)F)F

Specifications

Density 1.8
Boiling Point 158°C to 161°C
Flash Point None
Refractive Index 1.3
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 664.09
Percent Purity 97%
Chemical Name or Material Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.