accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27652-89-7

IUPAC Name: (4-chlorophenyl)(pyridin-2-yl)methanol
Synonyms: (4-chlorophenyl)(pyridin-2-yl)methanol
Molecular Weight (g/mol): 219.67
Molecular Formula: C12H10ClNO
InChi Key: ZFUPOFQRQNJDNS-UHFFFAOYNA-N
SMILES: OC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results