accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 276694-16-7

BOOH3CCH3H3CH3COHNNH2

CAS RN 276694-16-7

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzohydrazide
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzohydrazide
Molecular Weight (g/mol): 262.12
Molecular Formula: C13H19BN2O3
InChi Key: GFZSUVMUIACQHK-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results