CAS RN 2767983-73-1
CAS RN 2767983-73-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Chem-CLIP control probe
Control probe for Chem-CLIP experiments