accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 276869-41-1

OOOOHOONH(S)CH3H3CCH3

CAS RN 276869-41-1

IUPAC Name: (2S)-8-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-8-oxooctanoic acid
Synonyms: (2S)-8-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-8-oxooctanoic acid
Molecular Weight (g/mol): 467.56
Molecular Formula: C27H33NO6
InChi Key: ULPCAXORIFXUMN-QHCPKHFHSA-N
SMILES: CC(C)(C)OC(=O)CCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (2)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)