accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NSNHSHN

CAS RN 2770-75-4

IUPAC Name: 6-amino-1,2,3,4-tetrahydro-1,3,5-triazine-2,4-dithione
Synonyms: 6-amino-1,3-dihydro-1,3,5-triazine-2,4-dithione
Molecular Weight (g/mol): 160.21
Molecular Formula: C3H4N4S2
InChi Key: QQLZTRHXUSFZOM-UHFFFAOYSA-N
SMILES: NC1=NC(=S)NC(=S)N1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results