accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2772-45-4

IUPAC Name: 2,4-bis(2-phenylpropan-2-yl)phenol
Synonyms: 2,4-bis(2-phenylpropan-2-yl)phenol
Molecular Weight (g/mol): 330.47
Molecular Formula: C24H26O
InChi Key: FMUYQRFTLHAARI-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=CC=C1)C1=CC=C(O)C(=C1)C(C)(C)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results