Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Bis(alpha,alpha-dimethylbenzyl)phenol 97.0+%, TCI America™
SDP

Catalog No. B277225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B277225G 25 g
B2772500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B277225G Supplier TCI America Supplier No. B277225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2772-45-4
Molecular Formula C24H26O
Molecular Weight (g/mol) 330.471
InChI Key FMUYQRFTLHAARI-UHFFFAOYSA-N
Synonym 2,4-Di-alpha-cumylphenol
PubChem CID 76013
IUPAC Name 2,4-bis(2-phenylpropan-2-yl)phenol
SMILES CC(C)(C1=CC=CC=C1)C2=CC(=C(C=C2)O)C(C)(C)C3=CC=CC=C3

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 221°C
Quantity 25 g
Formula Weight 330.47
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Bis(alpha,alpha-dimethylbenzyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.