accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)SOS

CAS RN 27761-02-0

IUPAC Name: (2R)-2-hydroxy-1,2-bis(thiophen-2-yl)ethan-1-one
Synonyms: (2R)-2-hydroxy-1,2-bis(thiophen-2-yl)ethanone
Molecular Weight (g/mol): 224.29
Molecular Formula: C10H8O2S2
InChi Key: OGWZIOZTPNLTCR-VIFPVBQESA-N
SMILES: O[C@H](C(=O)C1=CC=CS1)C1=CC=CS1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results