Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Thenoin, TCI America™
SDP

Catalog No. T17625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17625G Supplier TCI America Supplier No. T17625G

missing translation for 'validMainframeMsg'

Specifications

CAS 27761-02-0
Melting Point 109°C
Color Yellow
Molecular Formula C10H8O2S2
MDL Number MFCD00022505
Quantity 5 g
Synonym 2,2'-thenoin, 2-hydroxy-1,2-di-2-thienylethanone, 2-hydroxy-1,2-di 2-thienyl ethan-1-one, 2-hydroxy-1,2-bis thiophen-2-yl ethan-1-one, 2-hydroxy-1,2-di thiophen-2-yl ethanone, 2-hydroxy-1,2-bis thiophen-2-yl ethanone, maybridge1_002119, 1,2-dithien-2-ylhydroxyethanone, 2-hydroxy-1,2-dithien-2-ylethanone, alpha-hydroxy-2-thenyl 2-thienyl ketone
InChI Key OGWZIOZTPNLTCR-UHFFFAOYSA-N
SMILES C1=CSC(=C1)C(C(=O)C2=CC=CS2)O
IUPAC Name 2-hydroxy-1,2-dithiophen-2-ylethanone
Molecular Weight (g/mol) 224.292
PubChem CID 2734800
Formula Weight 224.29
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Thenoin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.