accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3(Z)

CAS RN 2777-58-4

IUPAC Name: methyl (6Z)-octadec-6-enoate
Synonyms: methyl (6Z)-octadec-6-enoate
Molecular Weight (g/mol): 296.5
Molecular Formula: C19H36O2
InChi Key: QRTVDKVXAFJVRU-YPKPFQOOSA-N
SMILES: CCCCCCCCCCC\C=C/CCCCC(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results