Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl cis-6-Octadecenoate 98.0+%, TCI America™
SDP

Catalog No. M2306500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2306500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2306500MG Supplier TCI America Supplier No. M2306500MG
Add to Cart
Add to Cart

Specifications

CAS 2777-58-4
Melting Point -1°C
Color Colorless
Boiling Point 160°C
Molecular Formula C19H36O2
Quantity 500 mg
MDL Number MFCD00056193
Synonym methyl petroselinate, methyl cis-6-octadecenoate, petroselinic acid methyl ester, unii-774w29lwkq, methyl z-octadec-6-enoate, methyl 6z-octadec-6-enoate, 6-octadecenoic acid, methyl ester, z, methyl z-6-octadecenoate, methyl 6z-6-octadecenoate #, cis-6-octadecenoic acid methyl ester
InChI Key QRTVDKVXAFJVRU-YPKPFQOOSA-N
SMILES CCCCCCCCCCCC=CCCCCC(=O)OC
IUPAC Name methyl (Z)-octadec-6-enoate
Molecular Weight (g/mol) 296.495
PubChem CID 5362717
Formula Weight 296.50
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl cis-6-Octadecenoate
Show More Show Less
EINECSNumber (2)-0798
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.