accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 277756-44-2

IUPAC Name: 1-(trifluoromethyl)cyclopentane-1-carboxylic acid
Synonyms: 1-(trifluoromethyl)cyclopentane-1-carboxylic acid
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H9F3O2
InChi Key: DGQRYPPBAJNZFZ-UHFFFAOYSA-N
SMILES: OC(=O)C1(CCCC1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)